Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RUN and FYVE domain containing 1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human RUFY1 protein.
Supplier: Abnova Corporation H00080230D01P
Description
Sequence: MMEERANLMHMMKLSIKVLLQSALSLGRSLDADHAPLQQFFVVMEHCLKHGLKVKKSFIGQNKSFFGPLELVEKLCPEASDIATSVRNLPELKTAVGRGRAWLYLALMQKKLADYLKVLIDNKHLLSEFYEPEALMMEEEGMVIVGLLVGLNVLDANLCLKGEDLDSQVGVIDFSLYLKDVQDLDGGKEHERITDVLDQKNYVEELNRHLSCTVGDLQTKIDGLEKTNSKLQEELSAATDRICSLQEEQQQLREQNELIRERSEKSVEITKQDTKVELETYKQTRQGLDEMYSDVWKQLKEEKKVRLELEKELELQIGMKTEMEIAMKLLEKDTHEKQDTLVALRQQLEEVKAINLQMFHKAQNAESSLQQKNEAITSFEGKTNQVMSSMKQMEERLQHSERARQGAEERSHKLQQELGGRIGALQLQLSQLHEQCSSLEKELKSEKEQRQALQRELQHEKDTSSLLRMELQQVEGLKKELRELQDEKAELQKICEEQEQALQEMGLHLSQSKLKMEDIKEVNQALKGHAWLKDDEATHCRQCEKEFSISRRKHHCRNCGHIFCNTCSSNELALPSYPKPVRVCDSCHTLLLQRCSSTASSpecifications
RUN and FYVE domain containing 1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human RUFY1 protein. | |
RUFY1 | |
FLJ22251/RABIP4/ZFYVE12 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_001040451.1 | |
RUFY1 | |
RUFY1 (NP_001035541.1, 1 a.a. ∼ 600 a.a) full-length human protein. | |
100 μg | |
Ubiquitin/Proteasome | |
80230 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction