Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RUVBL1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523451

Catalog No. PIPA523451


Add to cart

Description

Description

Rvbs have been implicated in human pathologies, including carcinogenesis. For example Rvb1 overexpresssion has been reported in colon carcinoma, and Rvb1 SUMOylation has been shown to increase the proliferation and growth of prostate cancer cells. Rvb1 may also play a role in autoimmunity as high levels of RUVB1 autoantibodies have been detected in the sera of patients with autoimmune diseases including polymyositis/dermatomyosistic and autoimmune hepatitis. The extensive involvement of Rvbs in DNA-associated processes, including their interaction with a wide range of transcription factors, suggests that Rvb dysregulation may be involved in a number of other pathological processes yet to be elucidated.
Specifications

Specifications

RUVBL1
Polyclonal
Unconjugated
RUVBL1
Liquid
RUVBL1
portion of amino acids 1-50 of human Rvb1
Protein A
RUO
Antibody
Polyclonal
Human
Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q9Y265
ECP54, INO80H, NMP238, PONTIN, Pontin52, RVB1, TIH1, TIP49, TIP49A, 49 kDa TATA box-binding protein-interacting protein, 49 kDa TBP-interacting protein, 54 kDa erythrocyte cytosolic protein, ECP-54, INO80 complex subunit H, NMP 238, RuvB (E coli homolog)-like 1, TAP54-alpha, TATA binding protein interacting protein 49 kDa, TIP60-associated protein 54-alpha, nuclear matrix protein 238, pontin 52, ruvB-like 1
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
8607
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit