Learn More
RuvB-like 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061300
Catalog No. 06-130-0MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Anti-RuvB-like 2 Antibody is a rabbit polyclonal antibody for detection of RuvB-like 2 also known as 51 kDa erythrocyte cytosolic protein, Reptin52, RuvB (E coli homolog)-like 2, TBP-interacting protein and has been validated in western blotting.
Specifications
Specifications
RuvB-like 2
Unconjugated
Affinity Purified
Rabbit
Affinity purified
2-8°C for 12 months
Polyclonal
Human, Murine, Rat
Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
ECP-51; TIP48; TAP54-beta; ECP51; INO80; TIH2; RVB2; Rvb2; TIP49b
KLH-conjugated linear peptide corresponding to human RuvB-like 2.
100μL
Primary
NP_006657