Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RUVBL1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524579

Catalog No. PIPA524579

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

RUVBL1 possesses single-stranded DNA-stimulated ATPase and ATP-dependent DNA helicase (3 ft. to 5 ft.) activity. It is a component of the NuA4 histone acetyltransferase complex which is involved in transcriptional activation of select genes principally by acetylation of nucleosomal histone H4 and H2A. This modification may both alter nucleosome - DNA interactions and promote interaction of the modified histones with other proteins which positively regulate transcription. This complex may be required for the activation of transcriptional programs associated with oncogene and proto-oncogene mediated growth induction, tumor suppressor mediated growth arrest and replicative senescence, apoptosis, and DNA repair. The NuA4 complex ATPase and helicase activities seem to be, at least in part, contributed by the association of RUVBL1 and RUVBL2 with EP400. NuA4 may also play a direct role in DNA repair when recruited to sites of DNA damage. RUVBL1 plays an essential role in oncogenic transformation by MYC and also modulates transcriptional activation by the LEF1/TCF1 -CTNNB1 complex. High levels of autoantibodies against RUVBL1 are detected in sera of patients with autoimmune diseases such as polymyositis/dermatomyosistis and autoimmune hepatitis.
Specifications

Specifications

RUVBL1
Polyclonal
PBS with 0.09% sodium azide
Q9Y265
ECP54, INO80H, NMP238, PONTIN, Pontin52, RVB1, TIH1, TIP49, TIP49A, 49 kDa TATA box-binding protein-interacting protein, 49 kDa TBP-interacting protein, 54 kDa erythrocyte cytosolic protein, ECP-54, INO80 complex subunit H, NMP 238, RuvB (E coli homolog)-like 1, TAP54-alpha, TATA binding protein interacting protein 49 kDa, TIP60-a
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
8607
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
RUVBL1
Liquid
RUVBL1
human RUVBL1 recombit protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit