Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RUVBL2 Rabbit anti-Canine, Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA124414

Catalog No. PA124414


Add to cart

Description

Description

PA1-24414 detects RuvB-Like 2 (RUVBL2) in human, mouse, rat, and dog samples. PA1-24414 has been successfully used in Western blot and immunohistichemistry (paraffin) procedures. The PA1-24414 immunogen is a synthetic peptide corresponding to a region of human RUVBL2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

RuvB-Like 2 (48-kDa TATA box-binding protein-interacting protein, Reptin 52, RUVBL2) is the second human homologue of the bacterial RuvB gene. Bacterial RuvB protein is a DNA helicase essential for homologous recombination and DNA double-strand break repair. Functional analysis showed that this protein has both ATPase and DNA helicase activities. This gene is physically linked to the CGB/LHB gene cluster on chromosome 19q13.3, and is very close (55 nt) to the LHB gene, in the opposite orientation.
Specifications

Specifications

RuvBL2
Polyclonal
Unconjugated
RUVBL2
Liquid
RUVBL2
Synthetic peptide corresponding to a region of human RUVBL2.
Protein A
RUO
Antibody
Polyclonal
Canine, Human, Mouse, Rat
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q9Y230, Q9WTM5
CGI-46, ECP51, RVB2, TIP48, TIP49B
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
10856, 20174, 292907, 476418
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit