Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AHCY Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA512311

Catalog No. PIPA512311

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine, porcine and non-human primate based on sequence homology.

S-adenosylhomocysteine hydrolase (AHCY) catalyzes the reversible hydrolysis of S-adenosylhomocysteine (AdoHcy) to adenosine (Ado) and L-homocysteine (Hcy). Thus, it regulates the intracellular S-adenosylhomocysteine (SAH) concentration thought to be important for transmethylation reactions. Deficiency in this protein is one of the different causes of hypermethioninemia. S-adenosylhomocysteine hydrolase belongs to the adenosylhomocysteinase family.
Specifications

Specifications

AHCY
Polyclonal
Unconjugated
AHCY
Liquid
AHCY
KLH conjugated synthetic peptide between 79-110 amino acids from the N-terminal region of human S adenosylhomocysteine hydrolase (ACHY)
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Immunohistochemistry, Western Blot
1.5 mg/mL
PBS with 0.09% sodium azide
P23526
3D-structure; Acetylation; Cytoplasm; Direct protein sequencing; Hydrolase; NAD; One-carbon metabolism; Polymorphism.
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
191
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit