Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A4 Rabbit anti-Human, Polyclonal, Invitrogen™

Catalog No. 01673720
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-673-720 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-673-720 Supplier Thermo Scientific Supplier No. PA138589

Rabbit Polyclonal Antibody

S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an alpha and beta chain whereas S100B is composed of two beta chains. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.

Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is tonsil or melanoma tissue.
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100A4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution 1:100
Gene S100A4
Gene Accession No. P26447
Gene Alias 18A2, 42A, CAPL, FSP1, MTS1, P9KA, PEL98, S100 calcium-binding protein A4 calcium protein, calvasculin, metastasin, murine placental homolog, fibroblast-specific protein-1, leukemia multidrug resistance associated protein, maligt transformation suppression 1, placental calcium-binding protein, pr
Gene Symbols S100A4
Host Species Rabbit
Immunogen Recombit protein encoding full length human S100A4.
Purification Method Antigen affinity chromatography
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6275
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.