Learn More
S100A2, clone: SH-L1, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA125006
Description
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may have a tumor suppressor function. Chromosomal rearrangements and altered expression of this gene have been implicated in breast cancer.
MA1-25006 detects S100 alpha 2 from canine, human, rat, bovine, feline, goat, lizard, and porcine samples.MA1-25006 has been successfully used in immunoprecipitation, and Western blot procedures.The MA1-25006 immunogen is purified, full length, native pig protein.Specifications
S100 alpha-2 | |
Monoclonal | |
Unconjugated | |
P10462, P29034 | |
S100A2 | |
Purified, full length, native pig protein. | |
RUO | |
6273 | |
Store at -20°C, Avoid Freeze/Thaw Cycles | |
Ascites |
ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
SH-L1 | |
S100A2 | |
RP11-49N14.8, CAN19, MGC111539, S100L | |
Mouse | |
100 μL | |
Primary | |
Bovine, Canine, Feline, Goat, Human, Porcine, Rat, Reptile | |
Antibody | |
IgG1 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.