Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100B Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA131162

Catalog No. PA131162


Add to cart

Description

Description

PA1-31162 detects S100 beta from mouse and human samples. PA1-31162 has been successfully used in immunohistochemistry, immunoprecipitation, and Western blot applications. Positive control of rat brain tissue lysate suggested. The PA1-31162 immunogen is full length native protein homodimer (purified) (Human). Store at 4°C short term. For extended storage, aliquot and store at -20°C or below. Avoid freeze/thaw cycles.

S100B is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells and are involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100B may function in neurite extension, proliferation of melanoma cells, stimulation of Ca2+ fluxes, inhibition of PKC-mediated phosphorylation, astrocytosis and axonal proliferation, and inhibition of microtubule assembly. Chromosomal rearrangements and altered expression of this gene have been implicated in several diseases, including Alzheimer ft.s disease, Down ft.s syndrome, epilepsy, amyotrophic lateral sclerosis, melanoma, and type I diabetes.
Specifications

Specifications

S100B
Polyclonal
Unconjugated
S100B
Liquid
S100B
Full length native protein homodimer (purified) (Human).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
0.65 mg/mL
PBS with BSA, proprietary stabilizer and 0.02% sodium azide; pH 7.4
P04271, P50114
S100 calcium-binding protein B, S-100 protein beta subunit, S-100 protein beta chain, S100B
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
6285, 20203
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit