Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA138585

Catalog No. 01-673-718


Add to cart

Description

Description

A suggested positive control is melanoma or schwannoma.

S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an alpha and beta chain whereas S100B is composed of two beta chains. Antibody to S100 stains Schwannomas, ependymomas, astrogliomas, almost all benign and maligt melanomas and their metastases. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.
Specifications

Specifications

S100A1
Polyclonal
PBS with 1% BSA and 0.1% sodium azide; pH 7.6
P23297
S100, S100-alpha, S100A, S-100 protein alpha chain, S-100 protein subunit alpha, S100 alpha, S100 calcium-binding protein A1, S100 protein, alpha polypeptide, protein S100-A1
Rabbit
IgG
1 mL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6271
Immunohistochemistry (Paraffin)
Unconjugated
S100A1
Liquid
S100A1
Recombit protein encoding human S100 protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit