Learn More
S100, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA516257
Catalog No. PIPA516257
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an alpha and beta chain whereas S100B is composed of two beta chains. Antibody to S100 stains Schwannomas, ependymomas, astrogliomas, almost all benign and malignant melanomas and their metastases. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.
PA5-16257 targets S100 Protein in ICC and IHC (P) applications and shows reactivity with Bovine, Chicken, Human, Marsupial, mouse, and Rat samples.The PA5-16257 immunogen is s100 protein purified from cow brain.
Specifications
Specifications
S100
Polyclonal
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
P02638, P02639, P04271, P23297, P50114, P56565, P04631, P35467
S-100 protein alpha chain, S-100 protein subunit alpha, S100 alpha, S100 calcium-binding protein A1, S100 protein, alpha polypeptide, protein S100-A1
Rabbit
IgG
500μL
4° C
Primary
100858258, 20193, 20203, 25742, 295214, 424038, 525716, 528735, 6271, 6285
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
S100B
Liquid
S100A1, S100B
S100 protein purified from cow brain
Purified
RUO
Antibody
Polyclonal
Bovine, Chicken, Human, Marsupial, Murine, Rat