Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A10 Rat anti-Mouse, Clone: 344015, Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MA524080

Catalog No. PIMA524080


Add to cart

Description

Description

In direct ELISAs and Western blots, no cross-reactivity with recombit mouse (rm) S100A4, rmS100A8, rmS100A9, recombit human (rh) S100B, or rhS100P is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in exocytosis and endocytosis.
Specifications

Specifications

S100A10
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
P08207
42C, ANX2L, ANX2LG, CAL1L, CLP11, Ca[1], GP11, P11, p10, S100 calcium binding protein A10 (annexin II ligand, calpactin I, light polypeptide (p11)), S100 calcium-binding protein A10 (annexin II ligand, calpactin I, light polypeptide (p11)), annexin II ligand, calpactin I, light polypeptide, calpactin I light chain
Rat
IgG2A
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
20194
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
344015
Unconjugated
S100a10
Lyophilized
S100A10
E. coli-derived recombit mouse S100A10 Met1-Lys97
Protein A/G
RUO
Antibody
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit