Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A10 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526100

Catalog No. PIPA526100


Add to cart

Description

Description

The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation.
Specifications

Specifications

S100A10
Polyclonal
PBS with 0.09% sodium azide
P60903
42C, ANX2L, ANX2LG, CAL1L, CLP11, Ca[1], GP11, P11, p10, S100 calcium binding protein A10 (annexin II ligand, calpactin I, light polypeptide (p11)), S100 calcium-binding protein A10 (annexin II ligand, calpactin I, light polypeptide (p11)), annexin II ligand, calpactin I, light polypeptide, calpactin I light chain
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6281
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
S100A10
Liquid
S100A10
KLH conjugated synthetic peptide between 12-40 amino acids from the central region of human S100A10
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit