Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A11 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA512412

Catalog No. PIPA512412

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

S100A11 is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. This protein may function in motility, invasion, and tubulin polymerization. Chromosomal rearrangements and altered expression of S100A11 gene have been implicated in tumor metastasis.
Specifications

Specifications

S100A11
Polyclonal
PBS with 0.09% sodium azide
P31949
Protein S100-A11; S100 calcium-binding protein A11; Protein S100C; Calgizzarin; S100A11; MLN70; S100C
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6282
Western Blot
Unconjugated
S100A11
Liquid
S100A11
KLH conjugated synthetic peptide between 1-30 amino acids from human S100A11
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit