Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A11 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521330

Catalog No. PIPA521330


Add to cart

Description

Description

PA5-21330 targets S100A11 in ICC, IF, IHC (P), and WB applications and shows reactivity with Human samples. The PA5-21330 immunogen is a synthetic peptide corresponding to a region within amino acids 8 and 102 of S100A11 (Uniprot IDNo.P31949). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

S100A11
Polyclonal
Unconjugated
S100A11
Liquid
S100A11
Synthetic peptide corresponding to a region within amino acids 8 and 102 of S100A11 (Uniprot IDNo.P31949)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P31949
MLN70, S100C, MLN 70, S100 calcium-binding protein A11 (calgizzarin), calgizzarin, metastatic lymph node gene 70 protein, protein S100-A11, protein S100-C
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
6282
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit