Learn More
anti-S100A12, Polyclonal, Abnova
Mouse Polyclonal
Manufacturer:  Abnova Corporation H00006283B01
Catalog No. 89-006-187
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein is proposed to be involved in specific calcium-dependent signal transduction pathways and its regulatory effect on cytoskeletal components may modulate various neutrophil activities.
Detection Antibody, Western Blotting
Specifications
Specifications
S100 calcium binding protein A12
Serum
S100A12 (NP_005612.1, 1 a.a. ∽92 a.a) full-length human protein.
RUO
Primary
Human
Unconjugated
Murine
50μL
Y
Polyclonal