Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100P Mouse anti-Human, Unlabeled, Clone: 16, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610306

 View more versions of this product

Catalog No. BDB610306


Add to cart

Description

Description

The S100 multigene family consists of a number of small dimeric molecules around 100 residues in length. The original members of this family, S100α and S100β, were initially identified in bovine brain. S100P is a novel member of this family which was isolated from human placenta. The S100 family consists of at least 10 members which show a cell-type-specific expression pattern. Most S100 proteins are believed to be involved in cytoplasmic Ca2+ control. S100P consists of 95 amino acids and shares about 50% sequence identity with S100α and S100α. Like other S100 proteins, S100P functions as a homodimer and binds two Ca2+ ions per monomer. Each S100P molecule contains both a low affinity and a high affinity Ca2+ binding site.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

S100P
16
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50μg
Cell Biology
Primary
Human
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.