Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100P Mouse anti-Human, Unlabeled, Clone: 16, BD

Mouse Monoclonal Antibody

$225.00 - $419.00

Specifications

Antigen S100P
Isotype IgG1
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB610306
View Documents
BD Biosciences
610306
50μg Each for $225.00
Add to cart
 
BDB610307
View Documents
BD Biosciences
610307
150μg Each for $419.00
Add to cart
 
Description

Description

The S100 multigene family consists of a number of small dimeric molecules around 100 residues in length. The original members of this family, S100α and S100β, were initially identified in bovine brain. S100P is a novel member of this family which was isolated from human placenta. The S100 family consists of at least 10 members which show a cell-type-specific expression pattern. Most S100 proteins are believed to be involved in cytoplasmic Ca2+ control. S100P consists of 95 amino acids and shares about 50% sequence identity with S100α and S100α. Like other S100 proteins, S100P functions as a homodimer and binds two Ca2+ ions per monomer. Each S100P molecule contains both a low affinity and a high affinity Ca2+ binding site.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

S100P
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Unconjugated
Cell Biology
Murine
Affinity Purified
Monoclonal
Store undiluted at -20°C.
IgG1
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Western Blot
RUO
16
250μg/mL

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit