Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100Z Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524124

Catalog No. PIPA524124

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Members of the S100 protein family contain 2 calcium-binding EF-hands and exhibit cell-type specific expression patterns. For additional background information on S100 proteins, see MIM 114085.
Specifications

Specifications

S100Z
Polyclonal
PBS with 0.09% sodium azide
Q8WXG8
Gm625, S100-zeta, S100 calcium binding protein, zeta, S100 calcium-binding protein Z, protein S100-Z
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
170591, 268686
Flow Cytometry, Western Blot
Unconjugated
S100Z
Liquid
S100Z
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human S100Z
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit