Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC13A5 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524675

Catalog No. PIPA524675

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

S13A5 is a tricarboxylate plasma transporter with a preference for citrate.
Specifications

Specifications

SLC13A5
Polyclonal
Unconjugated
SLC13A5
Liquid
SLC13A5
KLH conjugated synthetic peptide between 151-180 amino acids from the central region of human S13A5
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q86YT5, Q67BT3
NACT, Na(+)/citrate cotransporter, sodium-coupled citrate transporter, sodium-dependent citrate transporter, solute carrier family 13 member 5, solute carrier family 13, member 5
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
237831, 284111
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit