Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-S1P1/EDG-1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP224622

 View more versions of this product

Catalog No. NBP224622


Add to cart

Description

Description

S1P1/EDG-1 Polyclonal antibody specifically detects S1P1/EDG-1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

S1P1/EDG-1
0.5 mg/mL
Western Blot
P21453
CD363, CD363 antigen, CHEDG1, D1S3362, EDG-1, EDG1edg-1, Endothelial differentiation G-protein coupled receptor 1, endothelial differentiation, sphingolipid G-protein-coupled receptor, 1, FLJ58121, S1P receptor 1, S1P receptor Edg-1, s1p1, S1P1ECGF1, sphingosine 1-phosphate receptor 1, sphingosine 1-phosphate receptor EDG1, Sphingosine 1-phosphate receptor Edg-1, sphingosine-1-phosphate receptor 1
Rabbit
IgG
0.1 mg
Cancer, Cytoskeleton Markers, GPCR, Lipid and Metabolism, Neuroscience, Signal Transduction
Primary
1901
Human
Western Blot
Unconjugated
PBS, 0.05% BSA with 0.05% Sodium Azide
Purified
S1PR1
A portion of amino acids 340-400 of human EDG-1 was used as the immunogen for this antibody.
Protein A purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
The amino acid sequence used as immunogen is 100% homologous in human, and 90% homologous in mouse and rat.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only