Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC6A12 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524005

Catalog No. PIPA524005

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Transports betaine and GABA. May have a role in regulation of GABAergic transmission in the brain through the reuptake of GABA into presynaptic terminals, as well as in osmotic regulation.
Specifications

Specifications

SLC6A12
Polyclonal
Unconjugated
SLC6A12
Liquid
SLC6A12
KLH conjugated synthetic peptide between 9-38 amino acids from the N-terminal region of human S6A12
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
P48065
BGT-1, BGT1, GAT2, Na(+)/Cl(-) betaine/GABA transporter, gamma-aminobutyric acid transporter, sodium- and chloride-dependent betaine transporter
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6539
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit