Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p70 S6 Kinase Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA512726

Catalog No. PIPA512726

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine, mouse, rat and rabbit based on sequence homology.

The p70 S6 kinase (p70 Ribosomal Protein S6 Kinase, p70S6K) is a 70 kDa member of the ribosomal S6 kinase (RSK) family of serine/threonine kinases. p70 S6 kinase is predomitly localized in the cytoplasm, and is essential in growth factors regulated cell proliferation, pathways involving cell motility, such as metastases, the immune response, and tissue repair. p70 S6 kinase acts downstream of phosphoinositide (PI) 3-kinase, and its main physiological target is the S6 ribosomal protein, which is involved in upregulation of protein synthesis. The kinase activity of p70 S6 kinase leads to an increase in protein synthesis and cell proliferation. Amplification of the region of DNA encoding p70 S6 kinase and overexpression are seen in some breast cancer cell lines.
Specifications

Specifications

p70 S6 Kinase
Polyclonal
PBS with 0.09% sodium azide
P23443
Ribosomal protein S6 kinase beta-1; Ribosomal protein S6 kinase I; S6K; S6K1; 70 kDa ribosomal protein S6 kinase 1; p70 S6 kinase alpha; p70(S6K)-alpha; p70-S6K; P70S6K; p70-alpha; RPS6KB1; STK14A
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6198
Immunohistochemistry, Western Blot
Unconjugated
RPS6KB1
Liquid
RPS6KB1
KLH conjugated synthetic peptide between 425-454 amino acids from human S6K (RPS6KB1)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit