Learn More
Salmonella LPS Mouse anti-Bacteria, Clone: A99H, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA518258
Catalog No. PIMA518258
Add to cart
Description
Description
This antibody is specific for common LPS core of all Salmonellae O-serogroups tested: A, B, C1, C2, D, E1, E3, E4, F, G1 and G2. This does not cross-react with E. coli (mix), Klebsiella, Citrobacter, Pseudomonas, Yersinia, Shigella, Proteus or Legionella. Purity is >90%. Recommended pairs for ELISA: MA5-18258 (capture) and MA5-18259 or MA5-18260 (conjugated, detection).
The genus Salmonella is a member of the family Enterobacteriaceae. The genus is composed of Gram negative bacilli that are facultative and flagellated (motile). Salmonellae possess 3 major surface antigens; the H or flagellar antigen (phase 1 and 2), the O or somatic antigen (part of the LPS moiety) and the Vi or capsular antigen (referred to as K in other Enterobacteriaceae). Salmonellae also possess the LPS endotoxin characteristic of Gram negative bacteria. This LPS is composed of an O polysaccharide (O antigen) an R core and the endotoxic inner Lipid A. Endotoxins evoke fever and can activate complement, kinin and clotting factors.
Specifications
Specifications
Salmonella LPS
Monoclonal
100 μg/mL
PBS with 0.1% sodium azide; pH 7.2
Salmonellae
Salmonellae sp. native protein
Protein A
RUO
Antibody
Monoclonal
ELISA, Immunofluorescence
A99H
Unconjugated
Liquid
Mouse
IgG2a
100 μg
4° C
Primary
Bacteria