Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAMHD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$204.00 - $409.50
Specifications
Antigen | SAMHD1 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 1-5ug/ml∼, Immunohistochemistry-Paraffin 5ug/ml |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP227410SS
|
Novus Biologicals
NBP227410SS |
0.025 mg |
Each for $204.00
|
|
|||||
NBP227410
|
Novus Biologicals
NBP227410 |
0.1 mg |
Each for $409.50
|
|
|||||
Description
SAMHD1 Polyclonal specifically detects SAMHD1 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SAMHD1 | |
Western Blot 1-5ug/ml∼, Immunohistochemistry-Paraffin 5ug/ml | |
Polyclonal | |
Rabbit | |
Stem Cell Markers | |
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
25939 | |
This antibody was raised against SAMHD1 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat, Primate | |
DCIPMg11, Dendritic cell-derived IFNG-induced protein, EC 3.1.4.-, HDDC1, Monocyte protein 5, MOP5, MOP-5AGS5, SAM domain and HD domain 1, SAM domain and HD domain-containing protein 1, SBBI88 | |
SAMHD1 | |
IgG | |
Affinity Purified | |
The amino acid sequence used as immunogen for this antibody is 100% homologous in human, monkey, gibbon, macaque, rhesus monkey, orangutan, and chimpanzee, 94% in old World monkey, and new world monkey, 83% in cow, horse, elephant, pig, and pole-cat,78% in panda, dog, mole rat, opossum, and rabbit, 72% in Chinese hamster, rat, and frog, and 61% homologous in zebra finch. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title