Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAMSN1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525771

Catalog No. PIPA525771

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

SAMSN1 is a member of a novel gene family of putative adaptors and scaffold proteins containing SH3 and SAM domains .
Specifications

Specifications

SAMSN1
Polyclonal
PBS with 0.09% sodium azide
Q9NSI8
HACS1, NASH1, SASH2, SH3D6B, SLy2, SAM and SH3 domain containing 2, SAM domain, SH3 domain and nuclear localisation signals, 1, SAM domain, SH3 domain and nuclear localization signals protein 1, SAM domain-containing protein SAMSN-1, SH3-SAM adaptor protein, Src homology domain 3 (SH3)-containi
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
64092
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
SAMSN1
Liquid
SAMSN1
KLH conjugated synthetic peptide between 255-283 amino acids from the central region of human SAMSN1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit