Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAP130 Mouse anti-Human, Mouse, Rat, Clone: 1YO 1B10, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 041549
Description
Use Anti-SAP130 Antibody, clone 1YO 1B10 (Mouse Monoclonal Antibody) validated in western blot, immunocytochemistry to detect SAP130 also known as 130 kDa Sin3-associated polypeptide, SF3b130.Specifications
SAP130 | |
Monoclonal | |
Unconjugated | |
NP_598714.1 | |
Mouse | |
Unpurified | |
Primary | |
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG1 κ |
Immunocytochemistry, Western Blot | |
1YO 1B10 | |
Unpurified mouse monoclonal IgG1κ preservative-free ascites. | |
SAP130, Sf3b3, Kiaa0017 | |
Ovalbumin-conjugated linear peptide corresponding to mouse SAP130. | |
100 μL | |
Human, Mouse, Rat | |
Ascites |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only