Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAP130, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA131762

Catalog No. PIPA131762

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

SAP130 is a subunit of the histone deacetylase (see HDAC1; MIM 601241)-dependent SIN3A . corepressor complex (Fleischer et al., 2003 [PubMed 12724404]).

PA1-31762 detects SAP130 from human samples.PA1-31762 has been successfully used in Western blot applications.The PA1-31762 immunogen is: Peptide with sequence VSKKLEDIRTRYAF, from C terminus of the protein sequence according to NP_036558.
Specifications

Specifications

SAP130
Polyclonal
Unconjugated
SAP130
Purified
SAP130
Peptide with sequence VSKKLEDIRTRYAF, from C terminus of the protein sequence according to NP_036558.
IgG fraction
RUO
Antibody
Polyclonal
Human, Murine
Western Blot
0.5mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q9H0E3, Q8BIH0
FLJ12761
Goat
IgG
50μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
79595
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit