Learn More
anti-SAR1B, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Manufacturer:  Novus Biologicals NBP132725
Catalog No. NBP132725
Add to cart
Description
Description
SAR1B Polyclonal specifically detects SAR1B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
SAR1B
1.0mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Q9Y6B6
2310075M17Rik, ANDD, CMRD, EC 3.6.5, GTP-binding protein B, GTP-binding protein SAR1b, GTP-binding protein Sara, SAR1 homolog B (S. cerevisiae), SAR1a gene homolog (S. cerevisiae) 2, SAR1a gene homolog 2, SAR1a gene homolog 2 (S. cerevisiae), SARA2GTBPB, SARB
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse and Rat. Predicted cross-reactivity based on sequence identity: Chicken (95%), Pig (98%) and Bovine (98%).
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Affinity Purified
SAR1B
Full length human SAR1B Recombinant protein.
22 kDa
100 ul
Signal Transduction
Primary
51128
Human, Mouse, Rat