Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS Coronavirus Nucleocapsid Mouse anti-Virus, Clone: 18F629.1, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA141515

Catalog No. 01-676-620


Add to cart

Description

Description

It has recently been shown that SARS is caused by a human coronavirus. Human coronaviruses are the major cause of upper respiratory tract illness in humans, such as the common cold. Coronaviruses are positive-stranded RNA viruses, featuring the largest viral RNA genomes known to date (27-31 kb). The first step in coronavirus infection is binding of the viral spike protein, a 139-kDa protein, to certain receptors on host cells. The spike protein is the main surface antigen of the coronavirus. The most prominent protein in the culture supernatants infected with SARS virus is a 46 kDa nucleocapsid protein. This suggests that the nucleocapsid protein is a major immunogen that may be useful for early diagnostics.
Specifications

Specifications

SARS Coronavirus Nucleocapsid
Monoclonal
Unconjugated
Liquid
Mouse
IgM
200 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
Virus
Western Blot
18F629.1
PBS with 0.2% gelatin and 0.05% sodium azide
SARS Nucleocapsid
Human nucleocapsid protein.
Protein G
RUO
Antibody
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit