Learn More
SARS Coronavirus Nucleocapsid Mouse anti-Virus, Clone: Ncap11, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA141032
Catalog No. 01-676-445
Add to cart
Description
Description
It has recently been shown that SARS is caused by a human coronavirus. Human coronaviruses are the major cause of upper respiratory tract illness in humans, such as the common cold. Coronaviruses are positive-stranded RNA viruses, featuring the largest viral RNA genomes known to date (27-31 kb). The first step in coronavirus infection is binding of the viral spike protein, a 139-kDa protein, to certain receptors on host cells. The spike protein is the main surface antigen of the coronavirus. The most prominent protein in the culture supernatants infected with SARS virus is a 46 kDa nucleocapsid protein. This suggests that the nucleocapsid protein is a major immunogen that may be useful for early diagnostics.
Specifications
Specifications
SARS Coronavirus Nucleocapsid
Monoclonal
Unconjugated
Liquid
Mouse
IgG2b, kappa
200 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
Virus
Immunocytochemistry, Immunofluorescence, Western Blot
Ncap11
PBS with 0.2% gelatin and 0.05% sodium azide
SARS Nucleocapsid
Full-length recombit SARS nucleocapsid protein.
Protein G
RUO
Antibody
Monoclonal