Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS Spike Protein Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 15 publications
Supplier: Novus Biologicals NB10056578SS
Description
SARS Spike Protein Polyclonal specifically detects SARS Spike Protein in SARS-CoV, SARS-CoV-2 samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Sars Spike Protein | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
1489668 | |
SARS-CoV, SARS-CoV-2 | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
1.0 mg/ml | |
Western Blot 0.05 - 1.0 ug/mL, Simple Western 1:40 - 1:160, Immunohistochemistry reported in scientific literature (PMID 34357881), Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 16081529) | |
COVID-19 Spike, E2, E2 glycoprotein, Human coronavirus spike glycoprotein, Peplomer protein, S, S glycoprotein, SARS coronavirus S protein, SARS coronavirus Spike Protein, SARS CoV, SARS CoV S glycoprotein, SARS CoV Spike protein, SARSCoV, SARS-CoV S protein, SARS-COV Spike, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe acute respiratory syndrome spike glycoprotein, Severe acute respiratory syndrome virus spike glycoprotein, Spike glycoprotein, VGL2 | |
The antibody was developed by immunizing rabbits with a synthetic peptide corresponding to amino acids 1124-1140 (C-GNCDVVIGIVNNTVYDP) from the S (Spike glycoprotein)(Spike protein S2') for the Human SARS coronavirus (Genbank accession no. YP_009724390) | |
0.025 mg | |
Primary | |
Full length Spike protein transfected into UM92 cells was used as a positive control and an approximate 139 kDa band was observed. Dot Blot results using recombinant proteins for cross-reactivity testing revealed high reactivity to SARS-CoV-2 Spike Protein, 1000-1200 a.a. (NBP2-90973) and low/no reactivity towards MERS Spike 2 or H1N1 (NBP1-99041). | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction