Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514984

Catalog No. PIPA514984

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine based on sequence homology.

SARS2 catalyzes the attachment of serine to tRNA(Ser). It is also able to aminoacylate tRNA(Sec) with serine, to form the misacylated tRNA L-seryl-tRNA(Sec), which will be further converted into selenocysteinyl-tRNA(Sec).
Specifications

Specifications

SARS2
Polyclonal
PBS with 0.09% sodium azide
Q9NP81
Seryl-tRNA synthetase, mitochondrial [Precursor]; EC 6.1.1.11; Seryl-tRNA(Ser/Sec) synthetase; Serine--tRNA ligase; SerRSmt; SARS2; SARSM
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54938
Immunohistochemistry, Western Blot
Unconjugated
SARS2
Liquid
SARS2
KLH conjugated synthetic peptide between 160-188 amino acids from the N-terminal region of human SARS2
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit