Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-SASH1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP126651

Catalog No. NBP126651


Add to cart

Description

Description

SASH1 Polyclonal antibody specifically detects SASH1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Western Blot (Negative).
Specifications

Specifications

SASH1
1.0 mg/mL
Immunohistochemistry 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:250-1:1000, Immunoprecipitation 3-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:500-1:2000, Western Blot (Negative)
NP_056093.3
dJ323M4, dJ323M4.1, KIAA0790RP3-323M4.1, PEPE1, Proline-glutamate repeat-containing protein, SAM and SH3 domain containing 1,2500002E12Rik, SAM and SH3 domain-containing protein 1, SH3D6A
Rabbit
IgG
100 ul
Cell Cycle and Replication
Primary
23328
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
SASH1
The immunogen recognized by this antibody maps to a region between residue 750 and 800 of human SAM and SH3 domain containing 1 using the numbering given in entry NP_056093.3 (GeneID 23328).
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Human. Based on 100% sequence identity, this antibody is predicted to react with Gorilla and Northern white-cheeked gibbon.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only