Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SATB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA520534

Catalog No. PIPA520534


Add to cart

Description

Description

A suggested positive control is A-20 cell lysate. PA5-20534 can be used with blocking peptide PEP-0654.

Human special AT-rich sequence-binding protein-1 (SATB1) is a nuclear matrix/scaffold-associated region DNA-binding protein, predomitly expressed in the thymus and pre-B cells. Like its homolog SATB2, SATB1 selectively binds double-stranded, special AT-rich DNA sequences in which one strand exclusively consists of well-mixed A, T, and C nucleotides. SATB1 contains a dimerization domain that shares similarity with the PDZ motif, identified as an indispensable element for high-affinity binding of SATB1 to DNA. SATB1 constitutes a functional nuclear architecture that has a  ft.cage-like ft. protein distribution surrounding heterochromatin and regulates gene expression through chromatin remodeling/HDAC (histone deacetylase complex) and transcription factors recruitment. SATB1 functions as a  ft.genome organizer ft. essential for proper T-cell development. Recent studies show that SATB1 is necessary for breast cancer cells to become metastatic, and when ectopically expressed in non-metastatic cells, can induce invasive activity in vivo. At least two isoforms of SATB1 are known to exist.
Specifications

Specifications

SATB1
Polyclonal
Unconjugated
SATB1
Liquid
SATB1
A 15 amino acid peptide near the carboxy terminus of the human SATB1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q01826, Q60611
DNA-binding protein SATB1; special AT-rich sequence binding protein 1 (binds to nuclear matrix/scaffold-associating DNA); special AT-rich sequence-binding protein 1
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
20230, 316164, 6304
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit