Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEC31B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525528

Catalog No. PIPA525528

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a protein of unknown function. The protein has moderate similarity to rat VAP1 protein which is an endosomal membrane-associated protein, containing a putative Ca2+/calmodulin-dependent kinase II phosphorylation site.
Specifications

Specifications

SEC31B
Polyclonal
PBS with 0.09% sodium azide
Q9NQW1
SEC31B-1, SEC31L2, SEC31-like 2, SEC31-like protein 2, SEC31-related protein B, protein transport protein Sec31B, secretory pathway component Sec31B-1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
25956
Flow Cytometry, Western Blot
Unconjugated
SEC31B
Liquid
SEC31B
KLH conjugated synthetic peptide between 1076-1104 amino acids from the C-terminal region of human SC31B
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit