Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Scaffold attachment factor B2, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP136982

Catalog No. NBP136982


Add to cart

Description

Description

Scaffold attachment factor B2 Polyclonal antibody specifically detects Scaffold attachment factor B2 in Human samples. It is validated for Peptide ELISA.
Specifications

Specifications

Scaffold attachment factor B2
0.5 mg/mL
Peptide ELISA Detection limit 1:32000
NP_055464.1
KIAA0138SAF-B2, scaffold attachment factor B2
Goat
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is not expected to cross-react with SAFB1.
ELISA
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
SAFB2
Peptide with sequence C-GEHNQRLEEHQARA corresponding to internal region according to NP_055464.1.
Immunogen affinity purified
RUO
Primary
9667
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only