Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCAND2P Mouse anti-Human, Clone: 5F1, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA522042

Catalog No. PIMA522042


Add to cart

Description

Description

Peptide Sequence: MAVAVDQQIQ TPSVQDLQIV KLEEDSHWEQ EISLQGNYPG PETSCQSFWH FRYQEASRPR EA

The SCAN domain is a highly conserved, leucine-rich motif of approximately 60 aa originally found within a subfamily of zinc finger proteins. This gene belongs to a family of genes that encode an isolated SCAN domain, but no zinc finger motif. Functional studies have established that the SCAN box is a protein interaction domain that mediates both hetero- and homoprotein associations, and maybe involved in regulation of transcriptional activity. Multiple transcript variants which encode the same isoform but differ only in their 3 ft. UTRs, and another variant which encodes a distinct isoform have been described for this gene.
Specifications

Specifications

SCAND2P
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q9GZW5
SCAN domain containing 2 pseudogene, SCAND2,
Mouse
IgG1, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
54581
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
5F1
Unconjugated
SCAND2P
Liquid
SCAND2P
SCAND2P (NP_071333, 1 a.a. ∽ 62 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit