Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SKAP2 Goat anti-Human, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA518014

Catalog No. PIPA518014


Add to cart

Description

Description

This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology.

The protein encoded by this gene belongs to the src family kinases. This protein is similar to the src kinase associated phosphoprotein 1. It is an adaptor protein that is thought to play an essential role in the src signaling pathway in various cells. It inhibits PTK2B/RAFTK activity and regulates alpha-synuclein phosphorylation.
Specifications

Specifications

SKAP2
Polyclonal
Unconjugated
SKAP2
Liquid
SKAP2
Synthetic peptide sequence (GLVPKAYIMEMYDI) corresponding to the C-terminus amino acids of SKAP2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
O75563
SKAP2; SCAP2; PRAP; src family associated phosphoprotein 2; MGC10411; MGC33304; RA70; SAPS; SCAP2; SKAP-HOM; SKAP55R; Fyn-associated phosphoprotein SKAP55 homologue; Pyk2/RAFTK-associated protein; src family associated phosphoprotein 2; src kinase-associated phosphoprotein of 55-related protein; src-associated adaptor protein
Goat
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
8935
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit