Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCD-1 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP126393AF647
Description
SCD-1 Polyclonal specifically detects SCD-1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
SCD-1 | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
O00767 | |
SCD | |
Synthetic peptide made to an internal portion of human SCD (within residues 50-100). [Swiss-Prot# O00767] | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
FADS5, MSTP008, SCD1, SCDOS, Stearoyl-CoA desaturase | |
Rabbit | |
41 kDa | |
0.1 mL | |
Cancer, Diabetes Research, Lipid and Metabolism, Signal Transduction, Stem Cell Markers | |
6319 | |
Store at 4C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction