Learn More
alpha-ENaC Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00124W
Catalog No. PIOSR00124W
Add to cart
Description
Description
SCNN1 is highly expressed in kidney and lung tissues, detected at intermediate levels in pancreas and liver, and at low levels in heart and placenta. Isoform 1 and isoform 2 are predominate in all tissues. Expression of isoform 3, isoform 4 and isoform 5 are very low or not detectable, except in lung and heart tissues.
Nonvoltage-gated, amiloride-sensitive, sodium channels control fluid and electrolyte transport across epithelia in many organs. These channels are heteromeric complexes consisting of 3 subunits: alpha, beta, and gamma. This gene encodes the alpha subunit, and mutations in this gene have been associated with pseudohypoaldosteronism type 1 (PHA1), a rare salt wasting disease resulting from target organ unresponsiveness to mineralocorticoids. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifications
Specifications
alpha-ENaC
Polyclonal
whole serum with no preservative
P37088
Epithelial a(+) channel subunit alpha, alpha-ENaC, Nonvoltage-gated sodium channel 1 subunit alpha, alpha-NaCH, SCNN1, FLJ21883, ENaCalpha
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Immunohistochemistry
Unconjugated
SCNN1A
Lyophilized
SCNN1A
A synthetic peptide from the extracellular domain of human amiloride sensitive sodium channel subunit alpha
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
6337