Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCP3 Mouse, Unlabeled, Clone: 25, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611230

Catalog No. BDB611230


Add to cart

Description

Description

Division of haploid germ cells during meiosis involves the pairing and recombination of homologous chromosomes. The synaptonemal complex (SC) is a meiotic protein structure that enhances chromosome pairing and segregation, affects the number and distribution of crossovers, and produces functional chiasmata from crossovers. The SC contains a central element (CE) and two lateral elements (LE) that are connected by transverse filaments (TF). Among the various SC components are two mammalian LE proteins, SCP2 and SCP3 (Synaptonemal Complex Proteins 2 and 3). SCP3 is highly similar to two other mammalian proteins, Xlr and Xmr, that are expressed in B and T lymphocytes and meiotic cells, respectively, and to members of the IF (intermediate filament) protein family. The SCP3 C-terminal region contains multiple coiled-coil domains that mediate protein-protein interactions. Via these domains, SCP3 self-assembles into transversely striated homopolymeric fibers in somatic cells. Such assembly also requires the N-terminal nonhelical domain. This function indicates that SCP3 may serve as an LE molecular scaffold to which other proteins bind and participate in recombination.

Host Species: Mouse
Clone: 25
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Rat SCP3 aa. 1-109
Formula Weight [Chemical]: 34/37kDa

Immunofluorescence, Western Blotting

Specifications

Specifications

SCP3
25
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
Synaptonemal Complex Protein-3
Affinity Purified
Rat SCP3 aa. 1-109
50μg
Cell Biology
Primary
Canine, Chicken, Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit