Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-SCP3/SYCP3, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$119.60 - $314.00

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A synthetic peptide made to the N-terminal region of the mouse SCP3 protein. [UniProt# P70281]
Antigen SCP3/SYCP3
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB300230
View Documents
Novus Biologicals
NB300230
0.1 ml
Each for $314.00
Add to cart
 
NB300230SS
View Documents
Novus Biologicals
NB300230SS
0.025 ml
Each for $119.60
Add to cart
 
Description

Description

SCP3/SYCP3 Polyclonal antibody specifically detects SCP3/SYCP3 in Human, Mouse, Porcine, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Rabbit
A synthetic peptide made to the N-terminal region of the mouse SCP3 protein. [UniProt# P70281]
Polyclonal
Primary
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide
IgG
SCP3/SYCP3
Affinity Purified
RUO
50511
1 mg/mL
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit