Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCRN1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA525190
Description
This antibody is predicted to react with bovine and rat based on sequence homology.
This gene likely encodes a member of the secernin family of proteins. A similar protein in rat functions in regulation of exocytosis in mast cells. Alternatively spliced transcript variants have been described.Specifications
SCRN1 | |
Polyclonal | |
SCRN1 | |
SES1, secernin-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
69938, 9805 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry, Western Blot | |
Unconjugated | |
Q12765, Q9CZC8 | |
SCRN1 | |
KLH conjugated synthetic peptide between 10-37 amino acids from the N-terminal region of human SCRN1 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction