Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCYL2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513751

Catalog No. PIPA513751

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

SCYL2 is a component of AP2-containing clathrin coated structures at the plasma membrane or of endocytic coated vesicles. SCYL2 is a probable serine/threonine-protein kinase that phosphorylates, in vitro, the beta2-subunit of the plasma membrane adapter complex AP2 and other proteins in presence of poly-L-lysine. SCYL2 may also regulate clathrin-dependent trafficking between the TGN and/or the endosomal system.
Specifications

Specifications

SCYL2
Polyclonal
PBS with 0.09% sodium azide
Q6P3W7, Q8CFE4
SCY1-like protein 2; Coated vesicle-associated kinase of 104 kDa; SCYL2; CVAK104; KIAA1360;
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
213326, 55681
Immunohistochemistry, Western Blot
Unconjugated
SCYL2
Liquid
SCYL2
KLH conjugated synthetic peptide between 643-672 amino acids from the C-terminal region of human SCYL2
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit