Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-SDCCAG3, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP131567

Catalog No. NBP131567


Add to cart

Description

Description

SDCCAG3 Polyclonal antibody specifically detects SDCCAG3 in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

SDCCAG3
1.0mg/mL
Western Blot 1:500-1:3000
Affinity Purified
SDCCAG3
Recombinant protein encompassing a sequence within the center region of human SDCCAG3. The exact sequence is proprietary.
48 kDa
100 ul
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Predicted cross-reactivity based on sequence identity: Bovine (89%). Not yet tested in any other species.
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Antigen NY-CO-3, NY-CO-3, serologically defined colon cancer antigen 3
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
10807
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only