Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SDHA Rabbit anti-Human, Mouse, Porcine, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523085

Catalog No. PIPA523085

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The target sequence has 94% sequence homology with rat and 93% sequence homology with porcine.

SDHA is an important component in the citric acid cycle and respiratory chain. SDHA converts succinate to fumarate, transfers electrons in the respiratory chain, and ultimately transfers electrons to ubiquinone. SDHA is localized within the mitochondrial inner membrane. Mutations in the gene have been associated with Leigh syndrome and Huntington ft.s disease. However, SDHA also appears to have tumor suppressor effects.
Specifications

Specifications

SDHA
Polyclonal
Unconjugated
SDHA
Liquid
SDHA
A synthetic peptide made to an C-terminal region of the human SDHA protein (within residues 500-664).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Porcine
Immunocytochemistry, Immunofluorescence, Western Blot
0.2 mg/mL
PBS with 0.1% BSA, 50% glycerol and 0.05% sodium azide
P31040, Q8K2B3, Q0QF01
CMD1GG, FP, PGL5, SDH1, SDH2, SDHF, flavoprotein subunit of complex II, succinate dehydrogenase [ubiquinone] flavoprotein subunit, mitochondrial, succinate dehydrogenase complex flavoprotein subunit
Rabbit
IgG
50 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
6389, 66945, 780433
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit