Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SDPR Rabbit anti-Hamster, Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526931
Description
This gene encodes a calcium-independent phospholipid-binding protein whose expression increases in serum-starved cells. This protein is a substrate for protein kinase C (PKC) phosphorylation and recruits polymerase I and transcript release factor (PTRF) to caveolae. Removal of this protein causes caveolae loss and its over-expression results in caveolae deformation and membrane tubulation.
Specifications
SDPR | |
Polyclonal | |
CAVIN2 | |
CAVIN2, PS-p68, SDR, cavin-2, phosphatidylserine binding protein, phosphatidylserine-binding protein, serum deprivation-response protein | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
100772097, 8436 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry, Western Blot | |
Unconjugated | |
O95810 | |
Cavin2 | |
KLH conjugated synthetic peptide between 109-135 amino acids from the central region of human SDR | |
400 μL | |
Primary | |
Hamster, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction