Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SDS Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526052

Catalog No. PIPA526052

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

SDS encodes one of three enzymes that are involved in metabolizing serine and glycine. L-serine dehydratase converts L-serine to pyruvate and ammonia and requires pyridoxal phosphate as a cofactor. The encoded protein can also metabolize threonine to NH4+ and 2-ketobutyrate. The encoded protein is found predomitly in the liver.
Specifications

Specifications

SDS
Polyclonal
Unconjugated
SDS
Liquid
SDS
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human SDS
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Western Blot
0.28 mg/mL
PBS with 0.09% sodium azide
P20132
SDH, L-serine ammonia-lyase, L-serine deaminase, L-serine dehydratase, L-serine dehydratase/L-threonine deaminase, L-threonine dehydratase, TDH
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10993
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit