Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEZ6L2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524862

Catalog No. PIPA524862


Add to cart

Description

Description

This antibody is predicted to react with bovine and mouse based on sequence homology.

SE6L2 may contribute to specialized endoplasmic reticulum functions in neurons.
Specifications

Specifications

SEZ6L2
Polyclonal
Unconjugated
SEZ6L2
Liquid
SEZ6L2
KLH conjugated synthetic peptide between 879-907 amino acids from the C-terminal region of human SE6L2
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q6UXD5
PSK-1, seizure 6-like protein 2, type I transmembrane receptor (seizure-related protein)
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
26470
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit